Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277256.1 | NP_001264185.1 | 1, 2 | 674 | ||
NM_033524.3 | NP_277059.1 | 1, 2 | 674 | |||
GenBank mRNA | AB063495.1 | - | 1, 2 | 20 | ||
AK155065.1 | - | 1, 2 | 637 | |||
AK155070.1 | - | 1, 2 | 676 | |||
BC057874.1 | - | 1, 2 | 107 | 5319749 | ||
BC079606.1 | - | 1, 2 | 295 | 30355455 |