Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037722.2 | NP_001032811.2 | 9 | 930 | ||
NM_009614.2 | NP_033744.1 | 9 | 930 | |||
GenBank mRNA | AF006196.1 | - | 9 | 891 | ||
AK039384.1 | - | 9 | 910 | |||
AK048901.1 | - | 9 | 917 | |||
AK142520.1 | - | 9 | 912 | |||
AK149684.1 | - | 9 | 911 | |||
AK149796.1 | - | 903 | ||||
AK151804.1 | - | 9 | 866 | |||
AK152725.1 | - | 9 | 866 | |||
AK152954.1 | - | 9 | 867 | |||
AK152973.1 | - | 9 | 867 | |||
AK170101.1 | - | 9 | 896 | |||
AK170365.1 | - | 9 | 910 | |||
BC009132.1 | - | 9 | 879 | 3152945 | ||
BC057909.1 | - | 8 | 1072 | |||
EF506571.1 | - | 3 | 191 |
Gene Symbol: 4731419I09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132440.1 | - | 2 | 588 |