Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277266.1 | NP_001264195.1 | 20 | 2425 | ||
NM_001291871.1 | NP_001278800.1 | 19 | 2387 | |||
NM_009615.6 | NP_033745.4 | 19 | 2368 | |||
GenBank mRNA | AB021709.1 | - | 19 | 2329 | ||
AJ007365.1 | - | 19 | 2168 | |||
AK137693.1 | - | 19 | 2275 | |||
AK139471.1 | - | 19 | 2274 | |||
AK142441.1 | - | 19 | 2327 | |||
AK144161.1 | - | 20 | 2355 | |||
AK149288.1 | - | 19 | 2271 | |||
BC048792.1 | - | 19 | 2275 | |||
BC094655.1 | - | 19 | 2282 | 5705503 | ||
BC114999.1 | - | 19 | 2228 | 6506501 | ||
BC138416.1 | - | 19 | 2247 | 8861436 | ||
BC138420.1 | - | 19 | 2247 | 8861440 | ||
BC145270.1 | - | 19 | 2247 | 9053824 | ||
CX220689.1 | - | 2 | 52 | |||
U69613.1 | - | 19 | 2236 | |||
U69614.1 | - | 19 | 2343 |