Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277266.1 | NP_001264195.1 | 16 | 2139 | ||
NM_001291871.1 | NP_001278800.1 | 15 | 2101 | |||
NM_009615.6 | NP_033745.4 | 15 | 2082 | |||
GenBank mRNA | AB021709.1 | - | 15 | 2043 | ||
AJ007365.1 | - | 15 | 1882 | |||
AK137183.1 | - | 8 | 2291 | |||
AK137693.1 | - | 15 | 1989 | |||
AK139471.1 | - | 15 | 1988 | |||
AK142441.1 | - | 15 | 2041 | |||
AK144161.1 | - | 16 | 2069 | |||
AK149288.1 | - | 15 | 1985 | |||
BC026052.1 | - | 12 | 1621 | |||
BC048792.1 | - | 15 | 1989 | |||
BC094655.1 | - | 15 | 1996 | 5705503 | ||
BC114999.1 | - | 15 | 1942 | 6506501 | ||
BC138416.1 | - | 15 | 1961 | 8861436 | ||
BC138420.1 | - | 15 | 1961 | 8861440 | ||
BC145270.1 | - | 15 | 1961 | 9053824 | ||
U69613.1 | - | 15 | 1950 | |||
U69614.1 | - | 15 | 2034 |