Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009632.2 | NP_033762.1 | 16 | 1635 | ||
GenBank mRNA | AF072521.1 | - | 16 | 1610 | ||
AJ007780.1 | - | 16 | 1648 | |||
BC052000.1 | - | 15 | 2020 | |||
BC062150.1 | - | 16 | 1635 | 6333875 |