Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005077.1 | - | 6 | 888 | ||
AK039267.1 | - | 5 | 548 | |||
S72537.1 | - | 7 | 743 |