Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009673.2 | NP_033803.1 | 7 | 777 | ||
GenBank mRNA | AK152185.1 | - | 7 | 612 | ||
AK153388.1 | - | 7 | 558 | |||
AK153476.1 | - | 7 | 541 | |||
D63423.1 | - | 7 | 515 | |||
U29396.1 | - | 7 | 534 |