Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF068707.1 | - | 15 | 1529 | ||
AK052384.1 | - | 15 | 1464 | |||
AK147177.1 | - | 16 | 1568 | |||
AK151844.1 | - | 16 | 1560 | |||
BC058402.1 | - | 14 | 1574 | |||
BC080798.1 | - | 15 | 1477 | |||
BC108374.1 | - | 16 | 1555 | 4936738 |