Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013475.4 | NP_038503.4 | 2, 3 | 298 | ||
GenBank mRNA | AK010926.1 | - | 2, 3 | 286 | ||
BC011037.1 | - | 2, 3 | 314 | |||
BC019811.1 | - | 2, 3 | 286 | 5125770 | ||
BC053338.1 | - | 2, 3 | 274 | 5125963 | ||
BY143138.1 | - | 2, 3 | 298 | |||
D10056.1 | - | 2, 3 | 272 | |||
S70439.1 | - | 2, 3 | 282 |