Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009700.2 | NP_033830.2 | 5 | 950 | ||
GenBank mRNA | AF469168.1 | - | 5 | 1024 | ||
AF469169.1 | - | 5 | 1206 | |||
AK039777.1 | - | 5 | 953 | |||
AK045357.1 | - | 4 | 601 | |||
AY484964.1 | - | 5 | 956 | |||
AY484965.1 | - | 6 | 1140 | |||
AY484966.1 | - | 6 | 1086 | |||
AY728043.1 | - | 5 | 970 | |||
AY728044.1 | - | 6 | 1154 | |||
AY728045.1 | - | 5 | 1082 | |||
AY803942.1 | - | 6 | 1061 | |||
AY803943.1 | - | 7 | 1191 | |||
AY803944.1 | - | 8 | 1375 | |||
BC024526.1 | - | 5 | 896 | 4985265 | ||
CT010362.1 | - | 4 | 897 | |||
U33012.1 | - | 4 | 912 | |||
U48397.1 | - | 4 | 1032 | |||
U48398.1 | - | 4 | 903 | |||
U48399.1 | - | 6 | 1014 | |||
U88623.1 | - | 5 | 951 |