Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175087.4 | NP_780296.1 | 4 | 722 | ||
GenBank mRNA | AK082699.1 | - | 4 | 723 | ||
BC115586.1 | - | 4 | 723 | 40102369 | ||
BC115587.1 | - | 4 | 722 | |||
DQ464060.1 | - | 4 | 723 | |||
DQ826418.1 | - | 4 | 714 |