Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290788.1 | NP_001277717.1 | 31 | 3805 | ||
NM_009702.3 | NP_033832.2 | 31 | 3805 | |||
GenBank mRNA | AK082873.1 | - | 31 | 3747 | ||
AK122310.1 | - | 31 | 3793 | |||
AK145547.1 | - | 31 | 3790 | |||
BC042479.1 | - | 31 | 3785 | 5363276 |