Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007533.5 | NP_031559.3 | 8 | 1067 | ||
GenBank mRNA | AK002435.1 | - | 8 | 1054 | ||
AK154734.1 | - | 8 | 1057 | |||
BC003787.1 | - | 8 | 1079 | 3600452 |
Gene Symbol: B3gnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154220.1 | - | 3088 |