Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn1sw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007538.3 | NP_031564.1 | 1 | 305 | ||
GenBank mRNA | AF190670.1 | - | 1 | 267 | ||
BC026021.1 | - | 1 | 272 | 4511808 | ||
BC050002.1 | - | 1 | 223 | |||
BC058267.1 | - | 1 | 253 | 6489727 | ||
CK620498.1 | - | 1 | 305 | |||
U49720.1 | - | 1 | 207 |
Set Membership: |
![]() |