Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291061.1 | NP_001277990.1 | 8 | 2010 | ||
NM_009751.2 | NP_033881.2 | 8 | 1992 | |||
GenBank mRNA | AB003147.1 | - | 8 | 1780 | ||
AK053766.1 | - | 8 | 1763 | |||
AK143180.1 | - | 8 | 1763 | |||
BC057580.1 | - | 8 | 1992 | 6412634 |