Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112796.2 | NP_001106267.1 | 5 | 2353 | ||
NM_009753.4 | NP_033883.2 | 5 | 2353 | |||
GenBank mRNA | AK033407.1 | - | 2 | 970 | ||
AK045975.1 | - | 5 | 2272 | |||
AK051379.1 | - | 5 | 2355 | |||
AK082523.1 | - | 5 | 2282 | |||
AK220497.1 | - | 5 | 2321 | |||
BC016192.1 | - | 5 | 2056 | 4483840 | ||
U90029.1 | - | 2 | 1220 |