Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btn1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013483.3 | NP_038511.1 | 5 | 1010 | ||
GenBank mRNA | AK142596.1 | - | 5 | 917 | ||
AK145168.1 | - | 5 | 1011 | |||
BC011497.1 | - | 5 | 984 | 3583827 | ||
BC031459.1 | - | 4 | 908 | 3582574 | ||
S80642.1 | - | 3 | 736 |