Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159319.2 | NP_001152791.1 | 2 | 304 | ||
NM_001282977.1 | NP_001269906.1 | 2 | 304 | |||
NM_001282978.1 | NP_001269907.1 | 2 | 304 | |||
NM_031173.4 | NP_112450.1 | 2 | 304 | |||
NM_145121.3 | NP_660099.2 | 2 | 304 | |||
GenBank mRNA | AB685743.1 | - | 2 | 288 | ||
AF068898.1 | - | 2 | 80 | |||
AF322905.1 | - | 2 | 308 | |||
AK155193.1 | - | 2 | 305 | |||
AY094172.1 | - | 2 | 102 | |||
AY094173.1 | - | 2 | 102 | |||
BC077713.1 | - | 2 | 300 | 30288369 | ||
BY080419.1 | - | 2 | 305 |