Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009787.2 | NP_033917.2 | 5, 6 | 1015 | ||
GenBank mRNA | AK028292.1 | - | 5, 6 | 865 | ||
AK146288.1 | - | 5, 6 | 848 | |||
AK148801.1 | - | 5, 6 | 937 | |||
AK150566.1 | - | 5, 6 | 896 | |||
AK161534.1 | - | 5, 6 | 853 | |||
AK168447.1 | - | 5, 6 | 819 | |||
AK169387.1 | - | 5, 6 | 860 | |||
BC066857.1 | - | 4, 5 | 614 | |||
BC141078.1 | - | 5, 6 | 911 | 9055690 | ||
J05186.1 | - | 5, 6 | 898 | |||
Y00884.1 | - | 4, 5 | 721 |