Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009794.3 | NP_033924.2 | 5 | 829 | ||
GenBank mRNA | AF015038.1 | - | 5 | 775 | ||
AK052141.1 | - | 5 | 804 | |||
AK053832.1 | - | 5 | 2115 | |||
AK078787.1 | - | 2 | 640 | |||
AK150695.1 | - | 5 | 803 | |||
AK152094.1 | - | 5 | 803 | |||
AK152790.1 | - | 5 | 804 | |||
AK152994.1 | - | 5 | 804 | |||
AK154855.1 | - | 5 | 843 | |||
AK184788.1 | - | 226 | ||||
BC054726.1 | - | 5 | 829 | 5703563 | ||
D38117.1 | - | 5 | 870 | |||
Y10139.1 | - | 5 | 711 |