Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007623.2 | NP_031649.2 | 5 | 1567 | ||
GenBank mRNA | AK141647.1 | - | 1 | 338 | ||
AK157869.1 | - | 5 | 1543 | |||
BC035199.1 | - | 5 | 1567 | 5137505 | ||
X62537.1 | - | 5 | 1634 | |||
Y13274.1 | - | 5 | 1491 |