Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009840.3 | NP_033970.3 | 8 | 944 | ||
GenBank mRNA | AK010882.1 | - | 8 | 945 | ||
AK075711.1 | - | 8 | 925 | |||
AK075880.1 | - | 8 | 933 | |||
AK145661.1 | - | 8 | 928 | |||
AK145754.1 | - | 8 | 927 | |||
AK145918.1 | - | 8 | 946 | |||
AK147038.1 | - | 8 | 947 | |||
AK159969.1 | - | 8 | 936 | |||
AK168465.1 | - | 8 | 917 | |||
AK172867.1 | - | 8 | 915 | |||
AK191019.1 | - | 4 | 334 | |||
AK211232.1 | - | 4 | 334 | |||
AK214517.1 | - | 4 | 350 | |||
AK219341.1 | - | 4 | 334 | |||
BC009007.1 | - | 8 | 902 | 3582639 | ||
Z37164.1 | - | 8 | 948 |