Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111058.1 | NP_001104528.1 | 3 | 736 | ||
NM_021293.3 | NP_067268.1 | 3 | 736 | |||
GenBank mRNA | AK046303.1 | - | 5 | 959 | ||
AK089205.1 | - | 3 | 654 | |||
AK155745.1 | - | 3 | 666 | |||
AK170937.1 | - | 3 | 704 | |||
BC132379.1 | - | 3 | 736 | 40130656 | ||
S71345.1 | - | 3 | 667 | |||
S71403.1 | - | 3 | 667 |