Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 7, 8 | 1171 | ||
NM_001159556.1 | NP_001153028.1 | 7, 8 | 1031 | |||
NM_001159557.1 | NP_001153029.1 | 8, 9 | 1082 | |||
NM_001159558.1 | NP_001153030.1 | 8, 9 | 1198 | |||
NM_007643.4 | NP_031669.3 | 7, 8 | 1174 | |||
GenBank mRNA | AK052825.1 | - | 7, 8 | 975 | ||
AK128908.1 | - | 8, 9 | 975 | |||
AK150648.1 | - | 7, 8 | 995 | |||
AK150865.1 | - | 3, 4 | 369 | |||
AK151355.1 | - | 7, 8 | 1010 | |||
AK152642.1 | - | 2, 3 | 166 | |||
AK152904.1 | - | 5, 6 | 681 | |||
AK172162.1 | - | 8, 9 | 1020 | |||
BC010262.1 | - | 7, 8 | 983 | 3481681 | ||
L23108.1 | - | 7, 8 | 992 |