Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007649.4 | NP_031675.1 | 3, 4 | 679 | ||
GenBank mRNA | AB196820.1 | - | 3, 4 | 640 | ||
AB196821.1 | - | 3, 4 | 640 | |||
AB196822.1 | - | 3, 4 | 640 | |||
AB196823.1 | - | 3, 4 | 640 | |||
AK007741.1 | - | 3, 4 | 680 | |||
AK084984.1 | - | 3, 4 | 708 | |||
AK146173.1 | - | 3, 4 | 726 | |||
AK151601.1 | - | 3, 4 | 670 | |||
BC060977.1 | - | 3, 4 | 707 | 30296173 | ||
X17501.1 | - | 3, 4 | 690 | |||
X53526.1 | - | 3, 4 | 690 |