Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042580.1 | NP_001036045.1 | 3 | 387 | ||
NM_001282966.1 | NP_001269895.1 | 3 | 431 | |||
NM_007653.3 | NP_031679.1 | 3 | 346 | |||
GenBank mRNA | AF159427.1 | - | 3 | 267 | ||
AK028220.1 | - | 3 | 284 | |||
AK146618.1 | - | 3 | 334 | |||
AK150647.1 | - | 3 | 341 | |||
AK151702.1 | - | 3 | 312 | |||
AK151963.1 | - | 3 | 286 | |||
AK152072.1 | - | 3 | 308 | |||
AK152372.1 | - | 3 | 312 | |||
AK159875.1 | - | 3 | 309 | |||
AK163785.1 | - | 3 | 308 | |||
AK166543.1 | - | 3 | 307 | |||
AK166766.1 | - | 3 | 306 | |||
AK170344.1 | - | 3 | 341 | |||
AK178582.1 | - | 1 | 142 | |||
AK182243.1 | - | 1 | 142 | |||
AK188949.1 | - | 1 | 144 | |||
AK208438.1 | - | 1 | 142 | |||
AK210314.1 | - | 1 | 142 | |||
AK216882.1 | - | 1 | 142 | |||
BC008108.1 | - | 3 | 293 | 3157993 | ||
BC012212.1 | - | 3 | 293 | 3157993 | ||
BC083161.1 | - | 3 | 278 | 6530532 | ||
BP769648.1 | - | 5, 6 | 569 | |||
CF355016.1 | - | 3 | 510 | |||
CT010179.1 | - | 2 | 242 | |||
D16432.1 | - | 3 | 296 |