Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009886.2 | NP_034016.2 | 25 | 7708 | ||
GenBank mRNA | AF031572.1 | - | 25 | 7708 | ||
AK220170.1 | - | 24 | 3602 |
Set Membership: |
G-protein coupled receptor |