Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007691.5 | NP_031717.2 | 13 | 1526 | ||
GenBank mRNA | AF016583.1 | - | 12 | 1435 | ||
AK011258.1 | - | 13 | 1470 | |||
AK033179.1 | - | 13 | 1496 | |||
AK045336.1 | - | 13 | 1468 | |||
AK166675.1 | - | 13 | 1526 | |||
BC037613.1 | - | 12 | 1239 | |||
BC053394.1 | - | 13 | 1463 | |||
BC064445.1 | - | 12 | 1925 | |||
BC100386.1 | - | 12 | 1601 | 30481390 |