Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007694.4 | NP_031720.1 | 4 | 2313 | ||
GenBank mRNA | AK077487.1 | - | 4 | 1882 | ||
AK077489.1 | - | 4 | 1921 | |||
AK133629.1 | - | 4 | 1918 | |||
AK158857.1 | - | 4 | 2314 | |||
BC014736.1 | - | 4 | 1914 | 4511248 | ||
X51429.1 | - | 4 | 1909 | |||
X53028.1 | - | 4 | 1919 |