Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013807.2 | NP_038835.2 | 7 | 907 | ||
GenBank mRNA | AK156754.1 | - | 7 | 919 | ||
BC031180.1 | - | 7 | 899 | 4988360 | ||
BC063051.1 | - | 7 | 907 | 5718130 | ||
U21392.1 | - | 7 | 754 |