Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007726.3 | NP_031752.1 | 2 | 1366 | ||
GenBank mRNA | AF153345.1 | - | 1 | 777 | ||
AK139417.1 | - | 2 | 974 | |||
AY522554.1 | - | 2 | 1354 | |||
BC070447.1 | - | 2 | 1366 | 30619104 | ||
BC075644.1 | - | 6 | 1783 | |||
BC079564.1 | - | 1 | 850 | 30536039 | ||
CA327850.1 | - | 1 | 282 | |||
CB244710.1 | - | 1 | 391 | |||
U17985.1 | - | 1 | 796 | |||
U40709.1 | - | 1 | 777 |
Set Membership: |
![]() |