Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053770.1 | - | 3 | 364 | ||
AF080580.1 | - | 4 | 427 | |||
AF098949.1 | - | 4 | 493 | |||
AK137841.1 | - | 2 | 2458 | |||
AK158380.1 | - | 4 | 478 | |||
BC028276.1 | - | 2 | 169 | |||
BC038681.1 | - | 4 | 460 | 1380996 | ||
U81277.1 | - | 3 | 209 |