Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252525.1 | NP_001239454.1 | 9 | 1458 | ||
NM_001252526.1 | NP_001239455.1 | 9 | 1440 | |||
NM_007755.5 | NP_031781.1 | 9 | 1455 | |||
GenBank mRNA | AK077799.1 | - | 1 | 83 | ||
AK136088.1 | - | 9 | 1459 | |||
BC125476.1 | - | 9 | 1375 | 40129991 | ||
BC144948.1 | - | 9 | 1357 | |||
Y08260.1 | - | 9 | 1374 |