Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013495.2 | NP_038523.2 | 12, 13 | 1539 | ||
GenBank mRNA | AF017175.1 | - | 1427 | |||
AK077454.1 | - | 12, 13 | 1493 | |||
AK128925.1 | - | 12, 13 | 1493 | |||
AK133472.1 | - | 12, 13 | 1494 | |||
AK136487.1 | - | 12, 13 | 1493 | |||
AK147770.1 | - | 12, 13 | 1494 | |||
AK149688.1 | - | 12, 13 | 1490 | |||
BC038395.1 | - | 11, 12 | 1474 | 2649352 | ||
BC046383.1 | - | 11, 12 | 1474 | 4160676 | ||
BC054791.1 | - | 12, 13 | 1520 | 6832731 | ||
D49472.1 | - | 11, 12 | 1466 |