Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277219.1 | NP_001264148.1 | 1 | 310 | ||
NM_001277221.1 | NP_001264150.1 | 1 | 310 | |||
NM_133656.5 | NP_598417.2 | 1 | 310 | |||
GenBank mRNA | AF239673.1 | - | 1 | 180 | ||
AK028179.1 | - | 1 | 287 | |||
AK028488.1 | - | 1 | 260 | |||
AK036905.1 | - | 1 | 261 | |||
AK052795.1 | - | 1 | 269 | |||
AK088519.1 | - | 1 | 281 | |||
AK139089.1 | - | 1 | 266 | |||
AK152549.1 | - | 1 | 283 | |||
AK154987.1 | - | 1 | 295 | |||
AK161356.1 | - | 1 | 260 | |||
AK161761.1 | - | 1 | 252 | |||
AK163291.1 | - | 1 | 285 | |||
BC012216.1 | - | 1 | 246 | 3992341 | ||
BC031149.1 | - | 1 | 243 | 4975654 | ||
S72408.1 | - | 1 | 235 |