Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136058.2 | NP_001129530.1 | 3 | 638 | ||
NM_007765.4 | NP_031791.3 | 3 | 503 | |||
GenBank mRNA | AB006714.1 | - | 3 | 475 | ||
AF501323.1 | - | 3 | 756 | |||
AK158793.1 | - | 3 | 582 | |||
AK159049.1 | - | 3 | 447 | |||
BC031738.1 | - | 3 | 432 | 4505284 | ||
BC065046.1 | - | 3 | 616 | 5686818 | ||
CA321187.1 | - | 3 | 638 | |||
CX562443.1 | - | 3 | 504 | |||
U72875.1 | - | 3 | 248 | |||
Y09080.1 | - | 3 | 478 |