Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007767.3 | NP_031793.2 | 1 | 1866 | ||
GenBank mRNA | AY013764.1 | - | 1 | 1866 | ||
BC054813.1 | - | 2 | 1875 | |||
BC150815.1 | - | 1 | 1974 | 9087726 | ||
BC150816.1 | - | 1 | 1974 | 9087727 | ||
D86917.1 | - | 1 | 1992 |
Gene Symbol: Gm38666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301256.1 | NP_001288185.1 | 1 | 1991 |