Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007789.3 | NP_031815.2 | 7 | 1513 | ||
GenBank mRNA | BC065118.1 | - | 7 | 1479 | 6853253 | |
X84727.1 | - | 7 | 1475 |