Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007790.3 | NP_031816.2 | 19 | 2134 | ||
GenBank mRNA | AF047601.1 | - | 19 | 2061 | ||
AF141294.1 | - | 19 | 2084 | |||
AK143996.1 | - | 19 | 2123 | |||
BC036330.1 | - | 7 | 834 | |||
BC057345.1 | - | 6 | 659 | 6811961 | ||
BC062935.1 | - | 19 | 2107 | 6838083 | ||
Y15128.1 | - | 5 | 562 |