Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010020.3 | NP_034150.1 | 5 | 833 | ||
GenBank mRNA | AF109072.1 | - | 5 | 756 | ||
AF109391.1 | - | 4 | 681 | |||
AJ238309.1 | - | 4 | 689 | |||
AK032629.1 | - | 5 | 791 | |||
AK136885.1 | - | 5 | 778 | |||
BC054119.1 | - | 5 | 833 | 6588897 |