Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010020.3 | NP_034150.1 | 10 | 1442 | ||
GenBank mRNA | AF109072.1 | - | 10 | 1365 | ||
AF109391.1 | - | 9 | 1290 | |||
AJ238309.1 | - | 9 | 1298 | |||
AK032629.1 | - | 10 | 1400 | |||
AK136885.1 | - | 10 | 1387 | |||
BC054119.1 | - | 10 | 1442 | 6588897 |