Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Des | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010043.2 | NP_034173.1 | 3 | 828 | ||
GenBank mRNA | AK131922.1 | - | 3 | 788 | ||
AK132391.1 | - | 3 | 792 | |||
BC031760.1 | - | 3 | 776 | 4219280 | ||
L22550.1 | - | 3 | 785 |