Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109750.1 | NP_001103220.1 | 6 | 965 | ||
NM_010048.3 | NP_034178.2 | 6 | 974 | |||
GenBank mRNA | AK042786.1 | - | 6 | 896 | ||
AK088131.1 | - | 6 | 895 | |||
AK133880.1 | - | 3 | 263 | |||
AK141594.1 | - | 6 | 940 | |||
AK145495.1 | - | 6 | 891 | |||
AK153026.1 | - | 6 | 762 | |||
AK161022.1 | - | 1725 | ||||
AK161359.1 | - | 6 | 690 | |||
AK163872.1 | - | 6 | 927 | |||
AK163903.1 | - | 6 | 903 | |||
AK190458.1 | - | 1 | 24 | |||
AK194718.1 | - | 1 | 24 | |||
AK202091.1 | - | 1 | 24 | |||
BC060636.1 | - | 6 | 915 | 6849298 | ||
BC062978.1 | - | 6 | 869 | 30356000 | ||
D78641.1 | - | 6 | 899 | |||
X95480.1 | - | 6 | 840 |