Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153150.2 | NP_694790.1 | 6 | 724 | ||
GenBank mRNA | AK005070.1 | - | 6 | 654 | ||
AK159289.1 | - | 6 | 687 | |||
AK170191.1 | - | 6 | 688 | |||
AK188281.1 | - | 3 | 192 | |||
AK206906.1 | - | 3 | 191 | |||
AK211700.1 | - | 3 | 192 | |||
AK219123.1 | - | 3 | 191 | |||
BC037087.1 | - | 6 | 684 | 5124403 |