Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006664.3 | NP_001006665.1 | 3 | 676 | ||
NM_001291120.1 | NP_001278049.1 | 4 | 643 | |||
NM_001291121.1 | NP_001278050.1 | 4 | 643 | |||
NM_001291122.1 | NP_001278051.1 | 4 | 643 | |||
NM_001291123.1 | NP_001278052.1 | 4 | 643 | |||
NM_013510.4 | NP_038538.1 | 3 | 594 | |||
GenBank mRNA | AB370262.1 | - | 2 | 189 | ||
AF061283.1 | - | 3 | 408 | |||
AK044744.1 | - | 4 | 469 | |||
AK122251.1 | - | 3 | 274 | |||
AK147272.1 | - | 4 | 505 | |||
AK147611.1 | - | 4 | 644 | |||
AK154704.1 | - | 4 | 462 | |||
AK165479.1 | - | 4 | 500 | |||
AW046764.1 | - | 3 | 373 | |||
BC034751.1 | - | 4 | 440 | 4954660 | ||
BE864786.1 | - | 5 | 419 | |||
BE985208.1 | - | 3 | 373 |