Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007939.2 | NP_031965.2 | 3 | 653 | ||
GenBank mRNA | AK220393.1 | - | 1 | 137 | ||
U72207.1 | - | 3 | 662 |