Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133658.1 | NP_598419.1 | 10 | 1734 | ||
GenBank mRNA | AK151188.1 | - | 10 | 1739 | ||
AK152347.1 | - | 10 | 1739 | |||
AK152988.1 | - | 10 | 1746 | |||
AK154745.1 | - | 10 | 1740 | |||
AK160187.1 | - | 4 | 838 | |||
BC016595.1 | - | 10 | 1705 | 3153847 | ||
BC022956.1 | - | 9 | 1838 | |||
BC024446.1 | - | 1 | 80 | |||
BC026575.1 | - | 10 | 1716 | 4955015 | ||
S71186.1 | - | 10 | 1734 |