Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010169.3 | NP_034299.2 | 2 | 908 | ||
GenBank mRNA | AK085990.1 | - | 2 | 949 | ||
AK146194.1 | - | 2 | 935 | |||
AK159282.1 | - | 2 | 963 | |||
AK160032.1 | - | 2 | 963 | |||
BC031516.1 | - | 2 | 908 | 3962777 | ||
L03529.1 | - | 2 | 863 |