Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010195.2 | NP_034325.2 | 5 | 829 | ||
GenBank mRNA | AF110818.1 | - | 5 | 542 | ||
AK047873.1 | - | 5 | 811 | |||
AK132387.1 | - | 5 | 829 | |||
AK134981.1 | - | 5 | 810 |