Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008026.5 | NP_032052.1 | 5 | 880 | ||
GenBank mRNA | AK036655.1 | - | 5 | 824 | ||
AK042111.1 | - | 5 | 793 | |||
AK083900.1 | - | 2 | 131 | |||
AK154714.1 | - | 5 | 797 | |||
BC138791.1 | - | 5 | 707 | 8861813 | ||
BC138792.1 | - | 5 | 707 | 8861814 | ||
X59421.1 | - | 5 | 880 |